Learn More
Hach Company Rochelle Salt Solution, 29 mL DB
Supplier: Hach Company 172533
Potassium sodium tartrate solution. Used to mask hardness interference in some analytical methods. 29 mL Dropping Bottle