Learn More
Hach Company Rochelle Salt-PVA Reagent, 100 mL MDB
Supplier:  Hach Company 2199432
Encompass_Preferred
Potassium sodium tartrate/polyvinyl alcohol solution. For determination of ammonia nitrogen and Total Kjeldahl Nitrogen by the Nessler method.
Catalog No. 50-199-5554
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.