Learn More
STEMCELL Technologies RoboSep Rat B Cell Isolation Kit, For processing: 1 x 10 9 cells
Supplier: STEMCELL Technologies 19644RF
The EasySep Rat B Cell Isolation Kit is designed to isolate B cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection. The EasySep procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep magnet and simply pouring or pipetting the desired cells into a new tube.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.