Learn More
STEMCELL Technologies RoboSep Non-Human Primate B Cell Isolation Kit, For processing: 1 x 10 9 cells
SDP
Supplier:  STEMCELL Technologies 1000347
Encompass
The EasySep Non-Human Primate B Cell Isolation Kit is designed to isolate untouched and highly purified B cells from fresh or previously frozen non-human primate peripheral blood mononuclear cells (PBMCs) in as little as 20 minutes by immunomagnetic negative selection. The EasySep procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep magnet and simply pouring or pipetting the desired cells into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry culture or DNA/RNA extraction.
Catalog No. 50-239-0363
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.