Learn More
STEMCELL Technologies RoboSep™ Mouse Pan-Naïve T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 19848RF
Encompass
The EasySep™ Mouse Pan-Naïve T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve T cells (CD11b, CD19, CD24, CD25, CD44, CD45R/B220, CD49b, TER119) and streptavidin-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.Click here (/easysep-15-minute-kits) to learn about our next-generation EasySep™ mouse cell isolation kits, featuring RapidSphere™ technology.
Catalog No. 50-197-4724
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.