Learn More
STEMCELL Technologies RoboSep Human CD45 Depletion Kit II, For processing: 2 x 10 9 cells
Supplier: STEMCELL Technologies 17898RF
The EasySep Human CD45 Depletion Kit II is designed to deplete CD45 cells from fresh or previously frozen human peripheral blood mononuclear cells. Unwanted cells are targeted with antibody complexes recognizing CD45 and magnetic particles. Labeled cells are separated using an EasySep magnet without the use of columns. Unwanted cells remain in the tube while desired cells are poured off. The CD45 antigen is expressed on all hematopoietic cells except erythrocytes and platelets.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.