Learn More
Enzo Life Sciences RIPA cell lysis buffer 2, (100ml)
Supplier: Enzo Life Sciences ADI801284
Formulation: Liquid. 50mM TRIS-hydrogen chloride, pH 7.4, containing 150mM sodium chloride, 1mM EDTA, 1mM EGTA, 1% Triton X-100, 1% sodium deoxycholate and 0.1% SDS. Long Term Storage: +4°C.