Learn More
Cytoskeleton, Inc. Rhodamine Phalloidin, 1x500 ul
Supplier: Cytoskeleton, Inc. PHDR1
Purity >99.5%, Phalloidin's usefulness as a laboratory tool is well established and lies in it's ability to inhibit microfilament de-polymerization. Thus, phalloidin stabilized microfilaments are used as substrates for the identification and characterization of the ever increasing number of microfilament associated proteins. In addition, rhodamine phalloidin can be used for staining the actin cytoskeleton in cells, and also as a fluorescent marker/stabilizer for microfilaments used in motility assays. The rhodamine phalloidin is provided as a lyophilized powder. When reconstituted with 500 µl methanol, it is at 14 µM (200 x stock).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.