Learn More
Perkin Elmer US LLC Rheflex Fingertight PEEK Fittings - 0.0625 in, Pkg. 10
Supplier: Perkin Elmer US LLC N9302676
Rheflex one-piece fingertight fittings are reusable and can be used with stainless steel, PEEK, or PTFE tubing. Quantity: Package of 10 Maximum Pressure: 5000 psi (350 bar) Tubing Size: 1/16 inch Thread Size: 10-32