Learn More
Proteintech Group Inc RFP-Trap Magnetic Particles M-270
SDP
Supplier:  Proteintech Group Inc RTD20
Encompass
RFP-Trap Magnetic Particles M-270 for immunoprecipitation (IP) of RFP-tagged proteins. RFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP. It consists of a RFP VHH/ Nanobody coupled to Magnetic Particles M-270.
Catalog No. 50-225-8755
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.