Learn More
Proteintech Group Inc RFP-Trap Magnetic Agarose, kit
SDP
Supplier:  Proteintech Group Inc RTMAK20
Encompass
The ChromoTek RFP-Trap Magnetic Agarose are affinity beads for IP of RFP-fusion proteins. It comprises a RFP Nanobody/ VHH coupled to magnetic agarose.
Catalog No. 50-225-8764
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.