Learn More
Proteintech Group Inc RFP-Booster ATTO594
Supplier: Proteintech Group Inc RBA59410
anti-RFP VHH/ Nanobody conjugated to fluorophore for IF, of mCherry and other red fluorescent protein-fusion proteins
This item is not returnable.
View return policy