Learn More
Electron Microscopy Sciences SEM Mount Holder Box Universal
Supplier:  Electron Microscopy Sciences 76520
Encompass_Preferred
SEM specimen mount holder box with insert and lid holds 12 mounts.
Use: 10 or 15 mm cylinders or 3.2 mm pins
Catalog No. 50-295-63
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.