Learn More
NEOGEN Reveal 3-D for Egg
SDP
Supplier:  NEOGEN 700002643
Encompass_Preferred
Reveal 3-D for Egg offers results in just 5 minutes and can be used in virtually any environment to screen environmental swabs and clean-in-place rinses. The Reveal 3-D line of food allergen tests kits is uniquely designed to guard against oversaturation and false negative results, with results displayed in three visual lines on the lateral flow test strip. A control line assures the test is working properly, and two other lines outline nondetectable, low, and high levels of egg.For more information visit https://www.neogen.com/solutions/Allergens/reveal-3d-egg
Catalog No. 50-201-5149
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.