Learn More
Electron Microscopy Sciences Anti-Vibration Platform Table Retractable Castor Feet
Supplier:  Electron Microscopy Sciences 6550
Encompass_Preferred
Incorporate a foot, which can be raised and lowered using a thumbwheel, allowing the workstation to be wheeled around and then securely mounted to the floor in a new location.
Catalog No. 50-344-63
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.