Learn More
Restek™ Tube and Pipe Fittings for GC Equipment
SDP
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Supplier:  Restek™ 21823
 View more versions of this product
Catalog No. 06-718-703
Description
Description
  • Available in brass and stainless steel
  • Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Note
Nuts and ferrules are not treated unless requested. Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Specifications
Specifications
GC Equipment
0.125 to 0.0625 in.
5/Pk.
Brass
Reducing Union Fitting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.