Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Stabilwax™-DA Capillary Columnsto be
Similar in polarity to DB-FFAP, OV-351, NUKOL FFAP, HP FFAP
Supplier: Restek™ 11008
Description
- Acidic functionality incorporated into the polymer structure
- Capable of analyzing acidic compounds without derivatization, significantly reducing adsorption of acids to increase sample capacity for volatile-free acids
- Ideal for analyzing distilled liquor and other alcoholic beverages
- Improved peak shapes for high molecular weight acids
- Compatible with some inorganic acids depending on the volatility
- Resistant to oxidative damage
- Equivalent to USP G25, G35 phases
- Do not rinse with water
Specifications
Stabilwax™-DA | |
0.25 mm | |
Free (Underivatized) Acids, and Various Inorganic Acids | |
0.1 μm | |
Polar | |
Resistant to oxidative damage |
250°C | |
G25, G35 | |
40°C to 250/260°C | |
30 m | |
Fused Silica |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction