Learn More
Restek™ Rtx™-200 Capillary Columns - 15m Length
General-purpose, fused silica columns for solvents, Freon fluorocarbons, alcohols, ketones, silanes, glycols, and drugs of abuse. Restek Rtx-200 Capillary Columns have excellent confirmation with Rtx-5 columns for phenols, nitrosamines, organochlorine pesticides, chlorinated hydrocarbons, and chlorophenoxy herbicides.
$463.00 - $642.00
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Temperature Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Temperature Range | Price | Quantity & Availability | |||
06-710-111
|
Restek™
15021 |
0.32 mm | 0.25 μm | -20°C to +320/340°C |
Each for $614.00
|
|
|||
06-710-148
|
Restek™
15052 |
0.53 mm | 1 μm | -20°C to +290/310°C |
Each for $586.00
|
|
|||
06-710-170
|
Restek™
15067119 |
0.53 mm | 1.5 µm | -20°C to +280/300°C |
Each for $624.00
|
|
|||
06-610-849
|
Restek™
15020 |
0.25 mm | 0.25 μm | -20°C to +320/340°C |
Each for $463.00
|
|
|||
06-610-856
|
Restek™
15035 |
0.25 mm | 0.5 μm | -20°C to +310/330°C |
Each for $501.00
|
|
|||
06-610-866
|
Restek™
15050 |
0.25 mm | 1 μm | -20°C to +290/310°C |
Each for $501.00
|
|
|||
06-610-857
|
Restek™
15036 |
0.32 mm | 0.5 μm | -20°C to +310/330°C |
Each for $568.00
|
|
|||
06-610-867
|
Restek™
15051 |
0.32 mm | 1 μm | -20°C to +290/310°C |
Each for $642.00
|
|
|||
06-610-850
|
Restek™
15022 |
0.53 mm | 0.25 μm | -20°C to +310/330°C |
Each for $586.00
|
|
|||
06-610-877
|
Restek™
15082 |
0.53 mm | 3 μm | -20°C to +260/280°C |
Each for $522.00
|
|
|||
06-710-101
|
Restek™
15007 |
0.53 mm | 0.1 μm | -20°C to +310/330°C | N/A | N/A | N/A | ||
06-610-873
|
Restek™
15067 |
0.53 mm | 1.5 μm | -20°C to +280/300°C | N/A | N/A | N/A | ||
Description
Rtx-200 columns have accomplished many difficult separations not possible on any other bonded stationary phase. The Rtx-200 column offers exceptional thermal stability, low bleed, and superior inertness, even for active compounds such as phenols, and with sensitive detectors such as ECDs, NPDs, and MSDs.
- Inert, midpolarity
- Crossbond trifluoropropylmethyl polysiloxane stationary phase has a unique selectivity that changes elution orders and resolves compounds that phenyl, cyano, or Carbowax phases cannot
- Equivalent to USP G6 phase
Order Info
Custom lengths and film thicknesses available.
Specifications
G6 | |
15 m | |
Fused Silica |