Learn More
Restek™ Ion Source Cleaning Powder
Clean surfaces that contact the sample or ion beam when encountering poor sensitivity and inadequate abundances at high masses
Supplier: Restek™ 22685
Description
- Aluminum oxide powder
- Size: 1 kg
Specifications
Cleaning Powder |