Learn More
Andwin Scientific REPLACEMENT POINT FOR SCRIBER
SDP
Supplier:  Andwin Scientific 52865118
Encompass
This replacement point is for the Diamond Scriber can be utilized for making permanent markings on several kinds of surfaces. including glass slides, test tubes, and laboratory glassware. This product features a clear aluminum handle that provides comfort for users. The tips are retractable for safe utilization.
Catalog No. NC9432884
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.