Learn More
Andwin Scientific REPLACEMENT BLADES SHARP/SHARP
SDP
Supplier:  Andwin Scientific 25609014
Encompass
The Tissue-Tek� Accu-Edge� replaceable Sharp/Sharp blade is an ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable.
Catalog No. NC9904856
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.