Learn More
Thermo Scientific™ Remel™ ImmunoWELL™ EBNA IgG
An EIA microwell test used for qualitative detection of IgG antibodies to Epstein-Barr Virus nuclear antigen-1 in serum.
Supplier:  Thermo Scientific™ R243230
Catalog No. 23-033-141
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Description
Description
Remel ImmunoWELL EBNA IgG is an EIA microwell test used for qualitative detection of IgG antibodies to Epstein-Barr Virus nuclear antigen-1 in serum.
Manufacturer: GenBio
Specifications
Specifications
ImmunoWELL™ EBNA IgG
Epstein-Barr Virus IgG
1 Kit
96 Wells
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.