Learn More
World Precision Instrument Reference Cells-RC2F
Supplier: World Precision Instrument RC2F
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.