Learn More
Biotium RedDot™1, 200X in water
Supplier: Biotium 400601
RedDot™1 is a far-red cell membrane-permeable nuclear dye similar to Draq5™. The dye is ideal for specifically staining the nuclei of live cells. RedDot™1 is highly thermostable and photostable, providing convenient handling for demanding applications such as confocal microscopy. The stain may also be used for DNA content analysis by flow cytometry. The dye has an excitation/emission of 662/694 nm when bound to DNA. Properties: Liquid; Store at 4°C or -20°C, protect from light; Ex/Em= 662/694 nm (with DNA).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.