Learn More
Neogen Diagnostics Red Stop Solution, 1 L, Microwell
Supplier:  Neogen Diagnostics 301476
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Red Stop Solution is a non-acidic, ready-to-use ELISA stop solution that contains no hazardous acids and can be used with our TMB substrates. Red Stop Solution does not increase background or optical density (OD) readings. Absorbance readings are taken at 650 nm within two hours of adding Red Stop Solution to the assay wells. Red Stop Solution offers a safer alternative to the traditional acids that are used to stop ELISA reactions. Stable for a minimum of 24-months at 2-8C or room temperature.
Catalog No. NC2215590