Learn More
Andwin Scientific RECTANGULAR TUB 15GAL EA
SDP
Supplier:  Andwin Scientific 674520
Encompass_Preferred
With a rugged caster frame, this rectangular mobile tub is able to store heavy solids or liquids of up to 60 liters. The tub and caster can easily be removed and cleaned. These products are approved by USDA and certified by NSF. A polyethylene cover is included.
Catalog No. 50-130-5848
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.