Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NT-4 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

$328.00 - $955.00

Specifications

Gene ID (Entrez) 4909
Molecular Weight (g/mol) 14 kDa
Name NT-4
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human NT-4 protein Gly81-Ala210
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Formulation Price Quantity & Availability  
268N4005
View Documents
R&D Systems™
268N4005
5 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Each for $328.00
Add to Cart
 
268N4005CF
View Documents
R&D Systems™
268N4005/CF
5 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Each for $328.00
Add to Cart
 
268N4025
View Documents
R&D Systems™
268N4025
25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Each for $955.00
Add to Cart
 
268N4025CF
View Documents
R&D Systems™
268N4025/CF
25 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Each for $955.00
Add to Cart
 
Description

Description

The Recombinant Human NT-4 Protein is derived from Sf 21 (baculovirus). The Recombinant Human NT-4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

4909
NT-4
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
14 kDa
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human NT-4 protein Gly81-Ala210
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.