Learn More
Proteintech Group Inc Recombinant Human IL-4 1000ug
SDP
Supplier:  Proteintech Group Inc HZ10041000UG
Encompass_Preferred
Animal-free Recombinant Human IL-4 also known as B cell-stimulatory factor-1. IL-4 is a monomeric, approximately 13 kDa and 19 kDa Th2 cytokine which shows pleiotropic effects during immune responses. Our IL-4 is produced in a human cell expression system with serum-free and chemically defined media. It has demonstrated greater stability under cell culture conditions making it ideal for efficient generation of Human Dendritic Cells.
Catalog No. 50-197-3838
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.