Learn More
Proteintech Group Inc Recombinant Human IL-28B 100ug
SDP
Supplier:  Proteintech Group Inc HZ1245100UG
Encompass_Preferred
Animal-free Recombinant Human IL-28B (IFN-lambda 3) is expressed in human 293 cells with an apparent molecular mass of 24 kDa. This cytokine is produced in a serum-free, chemically defined media. Production in human 293 cells offers authentic human post-translational modification which contributes to stability in cell growth media and other applications. The purity is greater than 95%.
Catalog No. 50-197-3942
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.