Learn More
Proteintech Group Inc Recombinant Human IL-2 10ug
SDP
Supplier:  Proteintech Group Inc HZ101510UG
Encompass_Preferred
Animal-free Recombinant Human IL-2 is an O-glycosylated, four alpha -helix bundle cytokine. It is expressed by gamma T cells, CD4+ and CD8+ T cells, eosinophils, B cells, and dendritic cells. IL-2 has potent stimulatory activity for antigen-activated T cells. It is a monomeric glycoprotein with an apparent molecular mass of 15 kDa. This cytokine is produced in a human cell expression system with serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation, which contributes to stability in cell growth media.
Catalog No. 50-197-3848
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.