Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human/Feline CXCL12/SDF-1 beta (aa 19-93)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$421.00
Specifications
Gene ID (Entrez) | 6387 |
---|---|
Molecular Weight (g/mol) | 8.8 kDa |
Name | CXCL12/SDF-1 beta aa 19-93 |
Quantity | 25 μg |
Source | E. coli-derived CXCL12/SDF-1 beta protein Ser19-Met93 |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
2716SD025
|
R&D Systems™
2716SD025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $421.00
|
|
|||||
2716SD025CF
|
R&D Systems™
2716SD025/CF |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
Each for $421.00
|
|
|||||
Description
The Recombinant Human/Feline CXCL12/SDF-1 beta (aa 19-93) is derived from E. coli. The Recombinant Human/Feline CXCL12/SDF-1 beta (aa 19-93) has been validated for the following applications: Bioactivity.Specifications
6387 | |
CXCL12/SDF-1 beta aa 19-93 | |
E. coli-derived CXCL12/SDF-1 beta protein Ser19-Met93 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
8.8 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title