Learn More
Proteintech Group Inc Recomb. Human pro IGF-II 100ug
SDP
Supplier:  Proteintech Group Inc HZ1161100UG
Encompass_Preferred
Animal-free Recombinant Human pro-IGF-II is expressed in human 293 cells as a monomeric glycoprotein with an apparent molecular mass of 25 kDa. Glycosylation contributes to stability in cell growth media and other applications. IGF-II expressed in E. coli is typically 7.5 kDa and nonglycosylated. This cytokine is produced in a human cell expression system with serum-free, chemically defined media.
Catalog No. 50-197-3914
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.