Learn More
Proteintech Group Inc Recomb. Human FGF-7 (KGF) 10ug
Supplier: Proteintech Group Inc HZ110010UG
Animal-free Recombinant Human FGF-7 (KGF) is expressed in human 293 cells as a monomeric glycoprotein with an apparent molecular mass of 17 and 30 kDa. FGF-7 appears to act on epithelial cells. It also seems to motivate and stimulate proliferation, migration, and differentiation of these cells. It plays a significant role in development, morphogenesis, wound healing, angiogenesis, and tumorigenesis. This cytokine is produced in a human cell expression system with serum-free, chemically defined media. Authentic glycosylation contributes to stability in cell growth media and other applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.