Learn More
Proteintech Group Inc Recomb. Hmn Thrombo. 10ug
Supplier: Proteintech Group Inc HZ124810UG
Animal-free Recombinant Human TPO is expressed from human 293 cells as a monomeric glycoprotein with an apparent molecular mass of 80 to 85 kDa. This cytokine is produced in a serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation, contributing to stability in cell growth media and other applications. C- terminal domain glycosylation is thought to be important for the secretion of TPO from cells and for survival of TPO in the circulation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.