Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Receiving Vessel
With Ground or Threaded Joints
Supplier: MilliporeSigma™ Supelco™ 64689U
Specifications
Receiving Vessel | |
Glass | |
Threaded |
2 mL | |
1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction