Learn More
Hach Company Cyanide Reagent Set, CyaniVer, 25 mL
Supplier: Hach Company 2242800
For determination of Cyanide by the Pyridine-Pyrazolone Method. Hach Method 8027. Range: 0.002 - 0.240 mg/L CN-. Sample Size: 25 mL Reagent Set Includes: CyaniVer 3, 4, and 5 Reagent Powder Pillows for 100 tests.