Learn More
Hach Company Chlorine (Free and Total) Reagent Set, DPD, 10 mL
Supplier: Hach Company 2459500
For determination of Chlorine (Free and Total) by the DPD Method. Hach Methods 8021 and 8167. Range: 0.02 - 2.00 mg/L Cl₂. Sample Size: 10 mL. Reagent Set contains DPD Free Chlorine Reagent and DPD Total Chlorine Reagent Powder Pillows for 100 tests each.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.