Learn More
Hach Company Acidity Reagent Set, Digital Titrator
Supplier:  Hach Company 2272800
Encompass_Preferred
For determination of acidity by Digital Titrator titration, with Methyl Orange (using bromphenol blue indicator) and Phenopthalein endpoints. Hach Methods 8201 (Methyl Orange) and 8202 (Phenolpthalein). Ranges: 10-160 and 1000-4000 mg/L as CaCO3. Reagent Set includes: Bromphenol Blue Powder Pillows, Phenolphthalein Powder Pillows and two Sodium Hydroxide Cartridges (0.1600 N and 1.600N). Approximately 40 - 100 tests
Catalog No. 50-199-5699
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.