Learn More
Research Products International Corp ReadyStrain Cell Straining Kits, 40µm, Blue, 50 per package
SDP
Supplier:  Research Products International Corp 401105
Encompass_Preferred
ReadyStrain Kits include cell strainer, 50ml tube and cap. Ideal for performing filtration of cell suspensions quickly and conveniently. All in one kit allows straining, processing of sample and storage. Sterile kit is ready to use with a strainer pre-attached to 50ml centrifuge tube. Ideal kit for opening under hood and reducing contamination risk. Supplied pre-assembled in individually wrapped easy to open "tear-strip' bags.
Includes strainer, 50ml tube and cap
Convenient sample processing
Sterile, pre-assembled
Catalog No. 50-197-7676
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.