Learn More
Metrohm USA REACTION VESSEL LG(250MM)100/C
SDP
Supplier:  Metrohm USA 61429050
Encompass
Reaction vessel for stability measurements involving volatile oils, mineral oils and biodiesel blends with the Biodiesel Rancimat. In combination with 6.2418.130. Set of 100 pieces.
Catalog No. NC9029162
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.