Learn More
Chemglass Life Sciences Reaction Block, 1 Dram (4mL) Vial, 15mm x 45mm, Gold, 48 Position, 19mm Hole Depth
Supplier: Chemglass Life Sciences CG199104
Reaction Block, 1 Dram (4mL) Vial, 15mm x 45mm, Gold, 48 Position, 19mm Hole Depth.Excellent heat transferWon't interfere with magnetic stirringHoles spaced for optimum stirring of all vialsColor-anodized for durability and easy identificationRound reaction blocks are designed to fit directly on 135mm diameter, round top hot plate stirrers. Aluminum construction offers excellent heat transfer without interfering with magnetic stirring. All blocks are anodized for increased strength, durability and chemical resistance. A thermowell is provided for insertion of a temperature sensor or contact thermometer. Available for scintillation vials and dram vials.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.