Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB780 Mouse Anti-Human CD39 (ENTPD1), BD Horizon™
Mouse Monoclonal Antibody
$168.00 - $412.00
Specifications
Antigen | CD39 (ENTPD1) |
---|---|
Clone | A1 (also known as A1/CD39) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB780 |
Description
CD39 (ENTPD1) Monoclonal antibody specifically detects CD39 (ENTPD1) in Human samples. It is validated for Flow cytometrySpecifications
CD39 (ENTPD1) | |
Flow Cytometry | |
RB780 | |
Mouse | |
Human | |
A1; ENTPD1; NTPDase-1; ecto-ATPDase 1; ecto-ATPase 1; ecto-apyrase | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
A1 (also known as A1/CD39) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
PHA-activated Human Peripheral Blood Mononuclear Cells | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title