Learn More
Andwin Scientific RAZOR BLADES 0.012IN 100PK
Supplier: Andwin Scientific 55411055
The single-edge industrial razor blades are machine-honed from hard carbon steel. The blades have thickness of 0.30 mm (0.012"). This product is sold in a pack of 100 individually wrapped blades.