Learn More
Andwin Scientific RAYON FILM FOR BIOLOGICAL CULT
SDP
Supplier:  Andwin Scientific 60941086
Encompass_Preferred
The Rayon Films are ideal for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth. The film is pierceable with tips or pipets for sample recovery, is non-cytotoxic, and is highly gas per
Catalog No. NC0605858
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.