Learn More
Innovative Research Inc Rat Vtrnctn Mltmrc Aff Pur
Supplier:  Innovative Research Inc IRTVTNMULTIAP100UG
Encompass_Preferred
Rat Vitronectin Multimeric Affinity Purified 100ug - Rat Vitronectin Multimeric Affinity Purified from Innovative Research has been purified from rat plasma by affinity chromatography. This is a Frozen liquid buffered in 0.05M Sodium Phosphate; 0.1M NaCl; pH 7.4 and a purity of >95% as determined by SDS-PAGE. More Details Species Rat Target Vitronectin Purification Method Affinity purified Purity >95% as determined by SDS-PAGE Analysis Source Rat plasma Storage Conditions -70 C Additional Information Prepared from plasma using urea as a denaturant. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including cell adhesion, cell differentiation, cell proliferation, protein-protein interactions, receptor binding, and tissue culture.
Catalog No. 50-274-3989
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.