Learn More
Biomatik Corporation Rat Transforming Growth Factor Beta 1 (TGFb1) CLIA Kit
Supplier:  Biomatik Corporation EKU08840
Encompass_Preferred
Rat Transforming Growth Factor Beta 1 (TGFb1) CLIA Kit for detecting TGFb1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.93pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P17246. Gene ID: 59086. Target Synonyms: TGF-B1; CED; DPD1; LAP; Camurati-Engelmann Disease; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6306
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.