Learn More
Cell Applications Inc RAT TAIL COLL COAT SOL 20ML
Supplier: Cell Applications Inc 12220
Enhances cell attachment and proliferation, or promotion of cell-specific morphology and function. Commonly used for endothelial cells, hepatocytes, muscle cells, and a variety of other cell types. 20 ml