Learn More
R&D Systems™ Rat SLC22A2/OCT2 (CAB52215) VersaClone cDNA
Catalog No. RDC0999 Shop All R&D Systems Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy
Solute carrier family 22 (organic cation transporter), member 2
The Rat SLC22A2/OCT2 (CAB52215) VersaClone cDNA clone contains the complete ORF for SLC22A2/OCT2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLC22A2/OCT2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Content And Storage
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
For Reasearch Use Only; Not for Use in Humans