Learn More
Sino Biological Rainin tip -200μL, Transparent, With filter, Box packing
Supplier:  Sino Biological PTM200YYNR
Encompass_Preferred
• Adapt to Rainin automation workstations and LTS manual pipette, major in used for liquid distribution and transfer.• Adapt to LTS platform.
Catalog No. 50-274-3838
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.