Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Rat Anti-TOX, BD Horizon™
Rat Monoclonal Antibody
$188.00 - $468.00
Specifications
Antigen | TOX |
---|---|
Clone | NAN448B |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
TOX Monoclonal antibody specifically detects TOX in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
TOX | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
9760, 252838 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
NAN448B | |
Flow Cytometry | |
R718 | |
Rat | |
Human, Mouse | |
TOX; TOX 1; TOX1 | |
Recombinant Human TOX Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title